accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 214907-33-2

BH3CH3CHOOH(E/Z)

CAS RN 214907-33-2

IUPAC Name: (4-methylpent-1-en-1-yl)boronic acid
Synonyms: 4-methylpent-1-en-1-ylboronic acid
Molecular Weight (g/mol): 127.98
Molecular Formula: C6H13BO2
InChi Key: OLTGERMANPOCTC-UHFFFAOYSA-N
SMILES: CC(C)CC=CB(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results