CAS RN 2150-37-0
IUPAC Name:
methyl 3,5-dimethoxybenzoate
Synonyms:
methyl 3,5-dimethoxybenzoate
Molecular Weight (g/mol):
196.2
Molecular Formula:
C10H12O4
InChi Key:
YXUIOVUOFQKWDM-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(OC)=CC(OC)=C1
1
–
1
of
1
results
1
Methyl 3,5-dimethoxybenzoate, 98%
CAS: 2150-37-0 Molecular Formula: C10H12O4 Molecular Weight (g/mol): 196.20 MDL Number: MFCD00008432 InChI Key: YXUIOVUOFQKWDM-UHFFFAOYSA-N Synonym: 3,5-dimethoxybenzoic acid methyl ester,benzoic acid, 3,5-dimethoxy-, methyl ester,3,5-dimethoxybenzoic acid methyl,methyl 3,5-bis methyloxy benzoate,acmc-1ccvs,rarechem al bf 0065,methyl 3,5-dimethoxybnezoate,ksc489q8p,ethyl 3,5-dimethoxybenzoate,methyl 3,5-dimethoxybenzoate PubChem CID: 75074 IUPAC Name: methyl 3,5-dimethoxybenzoate SMILES: COC(=O)C1=CC(OC)=CC(OC)=C1