accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2150-38-1

IUPAC Name: methyl 3,4-dimethoxybenzoate
Synonyms: methyl vanillate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: BIGQPYZPEWAPBG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)C(OC)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results