accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOCH3OCH3

CAS RN 2150-41-6

IUPAC Name: methyl 2,4-dimethoxybenzoate
Synonyms: methyl 2,4-dimethoxybenzoate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: IHIJFZWLGPEYPJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)C=C1OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results