accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2150-44-9

IUPAC Name: methyl 3,5-dihydroxybenzoate
Synonyms: methyl 3,5-dihydroxybenzoate
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: RNVFYQUEEMZKLR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=CC(O)=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results