accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 215309-01-6

IUPAC Name: 3-(4-methylpiperazin-1-yl)benzoic acid
Synonyms: 3-(4-methylpiperazin-1-yl)benzoic acid
Molecular Weight (g/mol): 220.27
Molecular Formula: C12H16N2O2
InChi Key: HALATUFUWLWCQV-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results