accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21535-97-7

IUPAC Name: 3-methyl-1-benzofuran
Synonyms: 3-methyl-benzofuran
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: ZRXHLJNBNWVNIM-UHFFFAOYSA-N
SMILES: CC1=COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results