accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21548-73-2

AgAgS

CAS RN 21548-73-2

IUPAC Name: disilver(1+) λ⁴-sulfanediidyl
Synonyms: silver sulfide
Molecular Weight (g/mol): 248.8
Molecular Formula: Ag2HS
InChi Key: WGXHXWWMANLXAQ-UHFFFAOYSA-N
SMILES: [SH--].[Ag+].[Ag+]
Physical Form 
  • (2)
  • (2)
  • (5)
Quantity 
  • (1)
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (2)
  • (5)
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)