accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCH3H2C

CAS RN 2155-94-4

IUPAC Name: dimethyl(prop-2-en-1-yl)amine
Synonyms: dimethyl(prop-2-en-1-yl)amine
Molecular Weight (g/mol): 85.15
Molecular Formula: C5H11N
InChi Key: GBCKRQRXNXQQPW-UHFFFAOYSA-N
SMILES: CN(C)CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results