accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2156-56-1

IUPAC Name: sodium 2,2-dichloroacetate
Synonyms: sodium dichloroacetate
Molecular Weight (g/mol): 150.92
Molecular Formula: C2HCl2NaO2
InChi Key: LUPNKHXLFSSUGS-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C(Cl)Cl
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results