accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2158-14-7

IUPAC Name: 4-acetamidobenzene-1-sulfonyl azide
Synonyms: 4-acetamidobenzenesulfonyl azide
Molecular Weight (g/mol): 240.24
Molecular Formula: C8H8N4O3S
InChi Key: NTMHWRHEGDRTPD-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results