accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 215868-81-8

IUPAC Name: (1-ethyl-1H-1,2,4-triazol-5-yl)methanol
Synonyms: (2-ethyl-1,2,4-triazol-3-yl)methanol
Molecular Weight (g/mol): 127.15
Molecular Formula: C5H9N3O
InChi Key: ZQYXCGFXZPLEKX-UHFFFAOYSA-N
SMILES: CCN1N=CN=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results