accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21594-52-5

IUPAC Name: 6-bromo-2H,3H-[1,3]oxazolo[4,5-b]pyridin-2-one
Synonyms: 6-bromo-3H-[1,3]oxazolo[4,5-b]pyridin-2-one
Molecular Weight (g/mol): 215.01
Molecular Formula: C6H3BrN2O2
InChi Key: VQPBRWIFFBIRRP-UHFFFAOYSA-N
SMILES: BrC1=CN=C2NC(=O)OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results