accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHHNOO

CAS RN 21598-06-1

IUPAC Name: 5-hydroxy-1H-indole-2-carboxylate
Synonyms: 5-hydroxy-1H-indole-2-carboxylate
Molecular Weight (g/mol): 176.15
Molecular Formula: C9H6NO3
InChi Key: BIMHWDJKNOMNLD-UHFFFAOYSA-M
SMILES: OC1=CC=C2NC(=CC2=C1)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results