accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2160-89-6

FFFOOCH2FFF

CAS RN 2160-89-6

IUPAC Name: 1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate
Synonyms: 1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate
Molecular Weight (g/mol): 222.09
Molecular Formula: C6H4F6O2
InChi Key: MNSWITGNWZSAMC-UHFFFAOYSA-N
SMILES: FC(F)(F)C(OC(=O)C=C)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results