accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 216019-28-2

IUPAC Name: [3-(propan-2-yl)phenyl]boronic acid
Synonyms: 3-isopropylphenylboronic acid
Molecular Weight (g/mol): 164.01
Molecular Formula: C9H13BO2
InChi Key: QSWLFBMVIGQONC-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results