accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SOONO

CAS RN 21626-70-0

IUPAC Name: 4-(morpholine-4-sulfonyl)aniline
Synonyms: 4-(morpholine-4-sulfonyl)aniline
Molecular Weight (g/mol): 242.29
Molecular Formula: C10H14N2O3S
InChi Key: FTKHPQFFQRKOJC-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(=O)(=O)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results