accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21635-88-1

IUPAC Name: oxetan-3-amine
Synonyms: oxetan-3-amine
Molecular Weight (g/mol): 73.1
Molecular Formula: C3H7NO
InChi Key: OJEOJUQOECNDND-UHFFFAOYSA-N
SMILES: NC1COC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results