accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 216393-56-5

IUPAC Name: 3-[(tert-butyldimethylsilyl)oxy]benzene-1-thiol
Synonyms: 3-[(tert-butyldimethylsilyl)oxy]benzenethiol
Molecular Weight (g/mol): 240.44
Molecular Formula: C12H20OSSi
InChi Key: KRHCTXFJCFMFOR-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC1=CC(S)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results