accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2164-33-2

ClOO

CAS RN 2164-33-2

IUPAC Name: 2-(chloromethyl)-2,3-dihydro-1,4-benzodioxine
Synonyms: 2-(chloromethyl)-2,3-dihydro-1,4-benzodioxine
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: AYPKYQSHFKQVDL-UHFFFAOYNA-N
SMILES: ClCC1COC2=CC=CC=C2O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results