accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONHNNOO

CAS RN 2164-83-2

IUPAC Name: 6-hydroxy-5-nitro-3,4-dihydropyrimidin-4-one
Synonyms: 6-hydroxy-5-nitro-3H-pyrimidin-4-one
Molecular Weight (g/mol): 157.09
Molecular Formula: C4H3N3O4
InChi Key: ABTLZAVJDRUDNG-UHFFFAOYSA-N
SMILES: OC1=C(C(=O)NC=N1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)