accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21653-40-7

IUPAC Name: hydrogen 5-[(diaminomethylidene)amino]-N-(4-nitrophenyl)-2-(phenylformamido)pentanamide chloride
Synonyms: hydrogen 5-[(diaminomethylidene)amino]-N-(4-nitrophenyl)-2-(phenylformamido)pentanamide chloride
Molecular Weight (g/mol): 434.88
Molecular Formula: C19H23ClN6O4
InChi Key: DEOKFPFLXFNAON-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)