accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21655-48-1

CH3NHHNH3C

CAS RN 21655-48-1

IUPAC Name: 2,6-dimethylpiperazine
Synonyms: 2,6-dimethylpiperazine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: IFNWESYYDINUHV-UHFFFAOYNA-N
SMILES: CC1CNCC(C)N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)