CAS RN 21663-61-6
IUPAC Name:
2,5-dichlorobenzonitrile
Synonyms:
2,5-dichlorobenzonitrile
Molecular Weight (g/mol):
172.01
Molecular Formula:
C7H3Cl2N
InChi Key:
LNGWRTKJZCBXGT-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(Cl)C(=C1)C#N
1
–
1
of
1
results
1
2,5-Dichlorobenzonitrile, 98%
CAS: 21663-61-6 Molecular Formula: C7H3Cl2N Molecular Weight (g/mol): 172.008 MDL Number: MFCD00019741 InChI Key: LNGWRTKJZCBXGT-UHFFFAOYSA-N Synonym: benzonitrile, 2,5-dichloro,2,5-dichloro-benzonitrile,2,5-dichlorobenzonitrite,2,5-dichlorobenzenecarbonitrile,pubchem3687,acmc-209fn5,ksc490q4h,2,5-dichlorobenzonitrile,attercop-chm at133057 PubChem CID: 89000 IUPAC Name: 2,5-dichlorobenzonitrile SMILES: C1=CC(=C(C=C1Cl)C#N)Cl