accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21674-96-4

CH3ONHSH2NN

CAS RN 21674-96-4

IUPAC Name: N-[4-(2-amino-1,3-thiazol-4-yl)phenyl]acetamide
Synonyms: N-[4-(2-amino-1,3-thiazol-4-yl)phenyl]acetamide
Molecular Weight (g/mol): 233.29
Molecular Formula: C11H11N3OS
InChi Key: VBBNSESFUHRMJU-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C1=CSC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results