accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SNNNOOH

CAS RN 216959-92-1

IUPAC Name: 4-methyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid
Synonyms: 4-methyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 221.23
Molecular Formula: C9H7N3O2S
InChi Key: XDZXQDHRHGIPRN-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=NC=CN=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results