accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21714-35-2

FFFNHSNH2Cl

CAS RN 21714-35-2

IUPAC Name: [2-chloro-5-(trifluoromethyl)phenyl]thiourea
Synonyms: 2-chloro-5-(trifluoromethyl)phenylthiourea
Molecular Weight (g/mol): 254.66
Molecular Formula: C8H6ClF3N2S
InChi Key: GCEQZLQDKSFFEF-UHFFFAOYSA-N
SMILES: NC(=S)NC1=C(Cl)C=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results