accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOOH

CAS RN 21715-90-2

IUPAC Name: 4-hydroxy-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Synonyms: 4-hydroxy-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: ZUSSTQCWRDLYJA-UHFFFAOYNA-N
SMILES: ON1C(=O)C2C3CC(C=C3)C2C1=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results