accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2172-12-5

CH3CH3HOCH3CH3HOCH3CH3HOY

CAS RN 2172-12-5

IUPAC Name: tris(propan-2-ol) yttrium
Synonyms: tris(isopropyl alcohol) yttrium
Molecular Weight (g/mol): 269.19
Molecular Formula: C9H24O3Y
InChi Key: NREVZTYRXVBFAQ-UHFFFAOYSA-N
SMILES: [Y].CC(C)O.CC(C)O.CC(C)O
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)