accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNO

CAS RN 21725-69-9

IUPAC Name: 2,3-dihydro-1,2-benzoxazol-3-one
Synonyms: 2H-1,2-benzoxazol-3-one
Molecular Weight (g/mol): 135.12
Molecular Formula: C7H5NO2
InChi Key: QLDQYRDCPNBPII-UHFFFAOYSA-N
SMILES: O=C1NOC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)