accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 217461-40-0

CH3NNONHO

CAS RN 217461-40-0

IUPAC Name: 1-[1-(cyclooctylmethyl)-3-(hydroxymethyl)piperidin-4-yl]-3-ethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-[1-(cyclooctylmethyl)-3-(hydroxymethyl)piperidin-4-yl]-3-ethyl-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 399.58
Molecular Formula: C24H37N3O2
InChi Key: MBGVUMXBUGIIBQ-UHFFFAOYNA-N
SMILES: CCN1C(=O)N(C2CCN(CC3CCCCCCC3)CC2CO)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results