accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21900-46-9

CH3H3COCl

CAS RN 21900-46-9

IUPAC Name: 2,3-dimethylbenzoyl chloride
Synonyms: 2,3-dimethylbenzoyl chloride
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: WFNKMVDATNLZBX-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(Cl)=O)=C1C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)