accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHg

CAS RN 21908-53-2

IUPAC Name: oxomercury
Synonyms: mercury(II) oxide
Molecular Weight (g/mol): 216.59
Molecular Formula: HgO
InChi Key: UKWHYYKOEPRTIC-UHFFFAOYSA-N
SMILES: O=[Hg]
Quantity 
  • (11)
  • (1)
  • (3)
  • (4)
  • (6)
Physical Form 
  • (15)
  • (2)
Grade 
  • (5)
  • (9)
  • (2)
Percent Purity 
  • (4)
  • (7)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (11)
  • (1)
  • (3)
  • (4)
  • (6)
  • (15)
  • (2)
  • (5)
  • (9)
  • (2)
  • (4)
  • (7)
  • (3)