accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONH2NH2N

CAS RN 219511-71-4

IUPAC Name: tert-butyl N-(diaminomethylidene)carbamate
Synonyms: tert-butyl N-(diaminomethylidene)carbamate
Molecular Weight (g/mol): 159.19
Molecular Formula: C6H13N3O2
InChi Key: UMOZLQVSOVNSCA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N=C(N)N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results