accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOClZrClClCl

CAS RN 21959-01-3

IUPAC Name: bis(oxolane); tetrachlorozirconium
Synonyms: bis(THF); zirconium(IV) chloride
Molecular Weight (g/mol): 377.24
Molecular Formula: C8H16Cl4O2Zr
InChi Key: VDJJKYYENYIHFF-UHFFFAOYSA-J
SMILES: Cl[Zr](Cl)(Cl)Cl.C1CCOC1.C1CCOC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)