accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHCH3H3C

CAS RN 2198-54-1

IUPAC Name: N-(3,4-dimethylphenyl)acetamide
Synonyms: 3,4-dimethylacetanilide
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: UAOIEEWQVAXCFY-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C)C(C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results