accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OCH3OCH3CH3O

CAS RN 21983-72-2

IUPAC Name: 3,3-dimethoxybutan-2-one
Synonyms: 3,3-dimethoxybutan-2-one
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: UFQBSPGKRRSATO-UHFFFAOYSA-N
SMILES: COC(C)(OC)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)