accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COONHCH3H3CH3C

CAS RN 2199-46-4

IUPAC Name: ethyl 3,4,5-trimethyl-1H-pyrrole-2-carboxylate
Synonyms: ethyl 3,4,5-trimethyl-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: WBOGFZDTCIQHSX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C(C)=C(C)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results