accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOBrOO

CAS RN 2199-87-3

IUPAC Name: 6-bromo-2-oxo-2H-chromene-3-carboxylate
Synonyms: 6-bromo-2-oxochromene-3-carboxylate
Molecular Weight (g/mol): 268.04
Molecular Formula: C10H4BrO4
InChi Key: XFQHPAXNKDYMOX-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=CC(Br)=CC=C2OC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results