accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2199-93-1

IUPAC Name: 3-acetyl-6-bromo-2H-chromen-2-one
Synonyms: 3-acetyl-6-bromochromen-2-one
Molecular Weight (g/mol): 267.08
Molecular Formula: C11H7BrO3
InChi Key: XFQYOFLFNKCHLG-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC(Br)=CC=C2OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results