accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22026-39-7

IUPAC Name: 2H-1,3-benzodioxole-5-carbohydrazide
Synonyms: 2H-1,3-benzodioxole-5-carbohydrazide
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: RAXBGBHBUFGWPG-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results