accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2203-80-7

IUPAC Name: 5-methylhex-1-yne
Synonyms: 1-hexyne, 5-methyl-
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: HKNANEMUCJGPMS-UHFFFAOYSA-N
SMILES: CC(C)CCC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)