accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrO

CAS RN 22037-28-1

IUPAC Name: 3-bromofuran
Synonyms: 3-bromofuran
Molecular Weight (g/mol): 146.97
Molecular Formula: C4H3BrO
InChi Key: LXWLEQZDXOQZGW-UHFFFAOYSA-N
SMILES: BrC1=COC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)