CAS RN 220442-79-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+)-15-Deoxyspergualin Trihydrochloride, TRC
CAS: 220442-79-5 Molecular Formula: C17H37N7O3 HCl Molecular Weight (g/mol): 496.9 Synonym: (+)-Gusperimus Trihydrochloride,(R)-7-[(Aminoiminomethyl)amino]-N-[2-[[4-[(3-aminopropyl)amino]butyl]amino]-1-hydroxy-2-oxoethyl]heptanamide Trihydrochloride IUPAC Name: N-[(1R)-2-[4-(3-aminopropylamino)butylamino]-1-hydroxy-2-oxoethyl]-7-carbamimidamidoheptanamide;trihydrochloride SMILES: Cl.Cl.Cl.NCCCNCCCCNC(=O)[C@@H](O)NC(=O)CCCCCCNC(=N)N