accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO(S)O

CAS RN 22047-88-7

IUPAC Name: (2S)-2-(benzyloxy)-2-phenylacetate
Synonyms: (2S)-2-(benzyloxy)-2-phenylacetate
Molecular Weight (g/mol): 241.27
Molecular Formula: C15H13O3
InChi Key: BGTPJDGURCMYML-AWEZNQCLSA-M
SMILES: [O-]C(=O)[C@@H](OCC1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results