accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 220498-02-2

IUPAC Name: (3S)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoate
Synonyms: (3S)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-phenylpropanoate
Molecular Weight (g/mol): 386.43
Molecular Formula: C24H20NO4
InChi Key: PTSLRPMRTOVHAB-QFIPXVFZSA-M
SMILES: [O-]C(=O)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results