CAS RN 220662-95-3
CAS RN 220662-95-3
IUPAC Name:
methyl[4-(pyridin-3-yl)but-3-en-1-yl]amine; oxalic acid
Synonyms:
oxalic acid; rivanicline
Molecular Weight (g/mol):
252.27
Molecular Formula:
C12H16N2O4
InChi Key:
WTIZFOAIQXMQHC-UHFFFAOYSA-N
SMILES:
OC(=O)C(O)=O.CNCCC=CC1=CN=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ RJR 2403 oxalate
CAS: 220662-95-3 Molecular Formula: C12H16N2O4