accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHSS

CAS RN 22068-49-1

IUPAC Name: 4-(1,3-dithiolan-2-yl)phenol
Synonyms: 4-(1,3-dithiolan-2-yl)phenol
Molecular Weight (g/mol): 198.3
Molecular Formula: C9H10OS2
InChi Key: LTNPCGWCUVDEEY-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1SCCS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results