accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2207-32-1

IUPAC Name: 5-chloro-2,1,3-benzothiadiazole
Synonyms: 5-chloro-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 170.61
Molecular Formula: C6H3ClN2S
InChi Key: VRNJWKISMWDTAY-UHFFFAOYSA-N
SMILES: ClC1=CC2=NSN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results