accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2207-75-2

IUPAC Name: potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate
Synonyms: potassium azaorotate
Molecular Weight (g/mol): 195.18
Molecular Formula: C4H2KN3O4
InChi Key: IAPCTXZQXAVYNG-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)C1=NC(=O)NC(=O)N1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)